siRNA Details
| Gene Symbol: LOC306766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014007.1 | NP_001014029.1 | 6, 7 | 735 | ||
| GenBank mRNA | AY325163.1 | - | 382 | |||
| BC085893.1 | - | 6, 7 | 735 | 7192798 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC306766| Gene Symbol: LOC306766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014007.1 | NP_001014029.1 | 6, 7 | 735 | ||
| GenBank mRNA | AY325163.1 | - | 382 | |||
| BC085893.1 | - | 6, 7 | 735 | 7192798 | ||