siRNA Details
| Gene Symbol: Ror2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107339.1 | NP_001100809.1 | 9 | 2212 | ||
| XM_008771495.1 | XP_008769717.1 | 8 | 1924 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ror2| Gene Symbol: Ror2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107339.1 | NP_001100809.1 | 9 | 2212 | ||
| XM_008771495.1 | XP_008769717.1 | 8 | 1924 | |||