siRNA Details
| Gene Symbol: Sema4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170563.1 | NP_001164034.1 | 11, 12 | 1847 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sema4d| Gene Symbol: Sema4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170563.1 | NP_001164034.1 | 11, 12 | 1847 | ||