siRNA Details
| Gene Symbol: Aspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014008.1 | NP_001014030.1 | 473 | |||
| XM_006253695.2 | XP_006253757.1 | 2343 | ||||
| GenBank mRNA | BC083711.1 | - | 473 | 7190620 | ||
| Gene Symbol: NEWGENE_1549776 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253707.2 | XP_006253769.1 | 3, 4 | 2343 | ||
| XM_008771557.1 | XP_008769779.1 | 2, 3 | 2419 | |||

Aspn
Genomic Map