siRNA Details
| Gene Symbol: Susd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107341.1 | NP_001100811.1 | 5 | 767 | ||
| XM_006253709.2 | XP_006253771.1 | 5 | 2343 | |||
| GenBank mRNA | FQ227785.1 | - | 5 | 952 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Susd3| Gene Symbol: Susd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107341.1 | NP_001100811.1 | 5 | 767 | ||
| XM_006253709.2 | XP_006253771.1 | 5 | 2343 | |||
| GenBank mRNA | FQ227785.1 | - | 5 | 952 | ||