siRNA Details
| Gene Symbol: Dek | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004255.1 | NP_001004255.1 | 9, 10 | 1154 | ||
| GenBank mRNA | BC079344.1 | - | 9, 10 | 1154 | 7107191 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dek| Gene Symbol: Dek | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004255.1 | NP_001004255.1 | 9, 10 | 1154 | ||
| GenBank mRNA | BC079344.1 | - | 9, 10 | 1154 | 7107191 | |