siRNA Details
| Gene Symbol: Gcnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001511.2 | NP_001001511.1 | 3 | 1522 | ||
| GenBank mRNA | AY435151.1 | - | 1042 | |||
| AY435152.1 | - | 1042 | ||||
| AY435153.1 | - | 3 | 1042 | |||
| BC099796.1 | - | 3 | 1522 | 7936188 | ||

Gcnt2
Genomic Map