siRNA Details
| Gene Symbol: Tfap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107345.1 | NP_001100815.1 | 5 | 1055 | ||
| XM_008771570.1 | XP_008769792.1 | 5 | 792 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tfap2a| Gene Symbol: Tfap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107345.1 | NP_001100815.1 | 5 | 1055 | ||
| XM_008771570.1 | XP_008769792.1 | 5 | 792 | |||