siRNA Details
| Gene Symbol: Tfap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107345.1 | NP_001100815.1 | 4 | 902 | ||
| XM_008771570.1 | XP_008769792.1 | 4 | 639 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001122948, NM_011547, U17285, U17286, AK013900, and others.

Tfap2a
Genomic Map