siRNA Details
| Gene Symbol: Cage1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012052.1 | NP_001012052.1 | 5 | 1410 | ||
| XM_006253808.2 | XP_006253870.1 | 5 | 1397 | |||
| XM_006253809.2 | XP_006253871.1 | 4 | 1088 | |||
| XM_006253810.2 | XP_006253872.1 | 5 | 1427 | |||
| XM_006253811.2 | XP_006253873.1 | 5 | 1511 | |||
| GenBank mRNA | BC081934.1 | - | 5 | 1410 | 7114574 | |
| FQ211932.1 | - | 5 | 1515 | |||

Cage1
Genomic Map