siRNA Details
| Gene Symbol: Mylk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253874.2 | XP_006253936.1 | 10 | 1982 | ||
| Gene Symbol: LOC103689985 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253865.2 | XP_006253927.1 | 9 | 1719 | ||
| XM_008771601.1 | XP_008769823.1 | 9 | 1719 | |||

Mylk4
Genomic Map