siRNA Details
| Gene Symbol: Fam65b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014009.2 | NP_001014031.2 | 17 | 1862 | ||
| XM_006253929.2 | XP_006253991.1 | 14 | 2291 | |||
| XM_006253930.1 | XP_006253992.1 | 14 | 1647 | |||
| XM_008771636.1 | XP_008769858.1 | 13 | 2005 | |||
| XM_008771637.1 | XP_008769859.1 | 13 | 1567 | |||
| XM_008771638.1 | XP_008769860.1 | 14 | 2132 | |||
| XM_008771639.1 | XP_008769861.1 | 13 | 1620 | |||
| GenBank mRNA | AY325194.1 | - | 17 | 1862 | ||

Fam65b
Genomic Map