siRNA Details
| Gene Symbol: Slc17a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107353.1 | NP_001100823.1 | 9 | 1413 | ||
| XM_006253966.2 | XP_006254028.1 | 9 | 1490 | |||
| XM_006253967.2 | XP_006254029.1 | 9 | 1237 | |||
| XM_006253968.2 | XP_006254030.1 | 9 | 1506 | |||
| XM_006253969.2 | XP_006254031.1 | 9 | 1490 | |||
| XM_008771654.1 | XP_008769876.1 | 9 | 2674 | |||
| XM_008771655.1 | XP_008769877.1 | 9 | 2421 | |||
| XM_008771656.1 | XP_008769878.1 | 9 | 2674 | |||

Slc17a2
Genomic Map