siRNA Details
| Gene Symbol: Slc17a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107353.1 | NP_001100823.1 | 11 | 1646 | ||
| XM_006253966.2 | XP_006254028.1 | 11 | 1723 | |||
| XM_006253967.2 | XP_006254029.1 | 11 | 1470 | |||
| XM_006253968.2 | XP_006254030.1 | 11 | 1739 | |||
| XM_006253969.2 | XP_006254031.1 | 11 | 1723 | |||
| XM_008771654.1 | XP_008769876.1 | 11 | 2907 | |||
| XM_008771655.1 | XP_008769877.1 | 11 | 2654 | |||
| XM_008771656.1 | XP_008769878.1 | 11 | 2907 | |||

Slc17a2
Genomic Map