siRNA Details
Gene Symbol: Zfp192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100574.1 | NP_001094044.1 | 5 | 1028 | ||
XM_008771718.1 | XP_008769940.1 | 5 | 1339 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100574.1 | NP_001094044.1 | 5 | 1028 | ||
XM_008771718.1 | XP_008769940.1 | 5 | 1339 |