siRNA Details
| Gene Symbol: Cdk13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271295.1 | NP_001258224.1 | 9 | 2975 | ||
| NM_001271296.1 | NP_001258225.1 | 9 | 2975 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk13| Gene Symbol: Cdk13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271295.1 | NP_001258224.1 | 9 | 2975 | ||
| NM_001271296.1 | NP_001258225.1 | 9 | 2975 | |||