siRNA Details
| Gene Symbol: Rab18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012468.1 | NP_001012486.1 | 5 | 330 | ||
| XM_006254132.2 | XP_006254194.1 | 5 | 387 | |||
| GenBank mRNA | BC089957.1 | - | 5 | 330 | 7312864 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab18| Gene Symbol: Rab18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012468.1 | NP_001012486.1 | 5 | 330 | ||
| XM_006254132.2 | XP_006254194.1 | 5 | 387 | |||
| GenBank mRNA | BC089957.1 | - | 5 | 330 | 7312864 | |