siRNA Details
| Gene Symbol: Wdr37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107362.1 | NP_001100832.1 | 3 | 419 | ||
| XM_008771733.1 | XP_008769955.1 | 2 | 768 | |||
| XM_008771734.1 | XP_008769956.1 | 2 | 288 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr37| Gene Symbol: Wdr37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107362.1 | NP_001100832.1 | 3 | 419 | ||
| XM_008771733.1 | XP_008769955.1 | 2 | 768 | |||
| XM_008771734.1 | XP_008769956.1 | 2 | 288 | |||