siRNA Details
| Gene Symbol: Net1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039023.1 | NP_001034112.1 | 2 | 234 | ||
| XM_006254206.2 | XP_006254268.1 | 4 | 452 | |||
| GenBank mRNA | BC100154.1 | - | 2 | 234 | 7460511 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Net1| Gene Symbol: Net1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039023.1 | NP_001034112.1 | 2 | 234 | ||
| XM_006254206.2 | XP_006254268.1 | 4 | 452 | |||
| GenBank mRNA | BC100154.1 | - | 2 | 234 | 7460511 | |