siRNA Details
| Gene Symbol: Nsun6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107367.2 | NP_001100837.1 | 8 | 1104 | ||
| XM_008771868.1 | XP_008770090.1 | 8 | 1513 | |||
| XM_008771869.1 | XP_008770091.1 | 8 | 1511 | |||
| XM_008771870.1 | XP_008770092.1 | 6 | 1251 | |||
| GenBank mRNA | BC162024.1 | - | 8 | 1104 | 9099401 | |

Nsun6
Genomic Map