siRNA Details
| Gene Symbol: Mastl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107369.1 | NP_001100839.1 | 157 | |||
| XM_006254340.2 | XP_006254402.1 | 219 | ||||
| XM_006254341.2 | XP_006254403.1 | 219 | ||||
| XM_006254342.2 | XP_006254404.1 | 223 | ||||
| GenBank mRNA | FQ224658.1 | - | 121 | |||
| Gene Symbol: LOC102553656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006254368.2 | XP_006254430.1 | 1 | 225 | ||

Mastl
Genomic Map