siRNA Details
| Gene Symbol: MGC114492 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024895.1 | NP_001020066.1 | 4 | 719 | ||
| XM_008773566.1 | XP_008771788.1 | 2 | 704 | |||
| GenBank mRNA | BC097440.1 | - | 4 | 719 | 7455831 | |
| Gene Symbol: RGD1559726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_225656.6 | XP_225656.1 | 1 | 735 | ||

MGC114492
Genomic Map