siRNA Details
| Gene Symbol: Neto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107371.1 | NP_001100841.1 | 4 | 1379 | ||
| XM_006255035.2 | XP_006255097.1 | 4 | 453 | |||
| XM_008772133.1 | XP_008770355.1 | 4 | 453 | |||
| XM_008772134.1 | XP_008770356.1 | 4 | 453 | |||

Neto1
Genomic Map