siRNA Details
| Gene Symbol: Timm21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008343.1 | NP_001008344.1 | 1 | 299 | ||
| GenBank mRNA | BC085823.1 | - | 1 | 299 | 7126835 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Timm21| Gene Symbol: Timm21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008343.1 | NP_001008344.1 | 1 | 299 | ||
| GenBank mRNA | BC085823.1 | - | 1 | 299 | 7126835 | |