siRNA Details
| Gene Symbol: Rbfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107373.1 | NP_001100843.1 | 5, 6 | 598 | ||
| XM_006254951.2 | XP_006255013.1 | 4, 5 | 703 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbfa| Gene Symbol: Rbfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107373.1 | NP_001100843.1 | 5, 6 | 598 | ||
| XM_006254951.2 | XP_006255013.1 | 4, 5 | 703 | |||