siRNA Details
| Gene Symbol: Me2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107376.2 | NP_001100846.2 | 15 | 1667 | ||
| XM_006254923.2 | XP_006254985.1 | 15 | 4814 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Me2| Gene Symbol: Me2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107376.2 | NP_001100846.2 | 15 | 1667 | ||
| XM_006254923.2 | XP_006254985.1 | 15 | 4814 | |||