siRNA Details
| Gene Symbol: Cep120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191697.1 | NP_001178626.1 | 7 | 830 | ||
| XM_008772141.1 | XP_008770363.1 | 6 | 1066 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep120| Gene Symbol: Cep120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191697.1 | NP_001178626.1 | 7 | 830 | ||
| XM_008772141.1 | XP_008770363.1 | 6 | 1066 | |||