siRNA Details
Gene Symbol: Sncaip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107379.1 | NP_001100849.1 | 10 | 2847 | ||
XM_006254719.2 | XP_006254781.1 | 10 | 2833 | |||
XM_006254720.2 | XP_006254782.1 | 10 | 2584 | |||
XM_006254721.2 | XP_006254783.1 | 10 | 2678 |
