siRNA Details
| Gene Symbol: Ablim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191698.1 | NP_001178627.1 | 14, 15 | 1723 | ||
| XM_008772144.1 | XP_008770366.1 | 13, 14 | 1693 | |||
| XM_008772145.1 | XP_008770367.1 | 11, 12 | 1507 | |||
| GenBank mRNA | FQ217083.1 | - | 7, 8 | 602 | ||

Ablim3
Genomic Map