siRNA Details
| Gene Symbol: Arhgef37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100974.1 | NP_001094444.1 | 4 | 472 | ||
| XM_006254807.2 | XP_006254869.1 | 4 | 531 | |||
| XM_006254808.2 | XP_006254870.1 | 4 | 2471 | |||
| XM_006254809.2 | XP_006254871.1 | 4 | 438 | |||
| XM_006254811.2 | XP_006254873.1 | 4 | 567 | |||
| GenBank mRNA | AB199794.1 | - | 4 | 472 | ||
| BC168236.1 | - | 514 | ||||

Arhgef37
Genomic Map