siRNA Details
| Gene Symbol: RGD1309362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024884.1 | NP_001020055.1 | 2 | 643 | ||
| XM_006254759.2 | XP_006254821.1 | 2 | 945 | |||
| GenBank mRNA | BC098065.1 | - | 2 | 643 | 7458735 | |
| Gene Symbol: LOC100910934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751792.3 | XP_003751840.2 | 1 | 643 | ||

RGD1309362
Genomic Map