siRNA Details
| Gene Symbol: Tcerg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107390.2 | NP_001100860.2 | 19, 20 | 2905 | ||
| XM_006254673.2 | XP_006254735.1 | 20, 21 | 2959 | |||
| XM_006254674.2 | XP_006254736.1 | 19, 20 | 2896 | |||
| XM_006254675.2 | XP_006254737.1 | 18, 19 | 2845 | |||
| GenBank mRNA | BC168862.1 | - | 19, 20 | 2865 | 7444403 | |

Tcerg1
Genomic Map