siRNA Details
| Gene Symbol: Diaph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107393.2 | NP_001100863.2 | 3 | 282 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Diaph1| Gene Symbol: Diaph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107393.2 | NP_001100863.2 | 3 | 282 | ||