siRNA Details
| Gene Symbol: Pcdhb22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001056235.5 | XP_001056235.1 | 1 | 2354 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhb22| Gene Symbol: Pcdhb22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001056235.5 | XP_001056235.1 | 1 | 2354 | ||