siRNA Details
| Gene Symbol: Hars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014012.2 | NP_001014034.2 | 11, 12 | 1443 | ||
| XM_006254575.2 | XP_006254637.1 | 10, 11 | 1353 | |||
| GenBank mRNA | BC089973.1 | - | 4, 5 | 607 | 7316402 | |

Hars2
Genomic Map