siRNA Details
Gene Symbol: Wdr55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017932.1 | NP_001017932.1 | 6 | 801 | ||
GenBank mRNA | BC127460.1 | - | 6 | 756 | 8367400 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wdr55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017932.1 | NP_001017932.1 | 6 | 801 | ||
GenBank mRNA | BC127460.1 | - | 6 | 756 | 8367400 |