siRNA Details
| Gene Symbol: Ctnna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007145.1 | NP_001007146.1 | 4 | 508 | ||
| GenBank mRNA | BC085789.1 | - | 4 | 508 | 7108824 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctnna1| Gene Symbol: Ctnna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007145.1 | NP_001007146.1 | 4 | 508 | ||
| GenBank mRNA | BC085789.1 | - | 4 | 508 | 7108824 | |