siRNA Details
| Gene Symbol: Ctnna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007145.1 | NP_001007146.1 | 3 | 354 | ||
| GenBank mRNA | BC085789.1 | - | 3 | 354 | 7108824 | |
| Gene Symbol: LOC100911326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772063.1 | XP_008770285.1 | 2 | 467 | ||

Ctnna1
Genomic Map