siRNA Details
| Gene Symbol: Cdc25c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107396.1 | NP_001100866.1 | 2 | 618 | ||
| XM_006254579.2 | XP_006254641.1 | 2 | 568 | |||
| XM_006254580.2 | XP_006254642.1 | 2 | 568 | |||
| XM_008772039.1 | XP_008770261.1 | 3 | 480 | |||

Cdc25c
Genomic Map