siRNA Details
| Gene Symbol: Epb41l4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107397.2 | NP_001100867.1 | 3 | 252 | ||
| XM_006254582.2 | XP_006254644.1 | 8 | 3524 | |||
| XM_006254583.2 | XP_006254645.1 | 8 | 2630 | |||
| XM_006254584.2 | XP_006254646.1 | 9 | 2276 | |||
| XM_006254588.2 | XP_006254650.1 | 8 | 830 | |||
| XM_008772040.1 | XP_008770262.1 | 8 | 607 | |||
| GenBank mRNA | BC168858.1 | - | 3 | 252 | 7097125 | |

Epb41l4a
Genomic Map