siRNA Details
| Gene Symbol: Ammecr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107399.1 | NP_001100869.1 | 5 | 853 | ||
| XM_006254590.2 | XP_006254652.1 | 5 | 1667 | |||
| XM_006254591.2 | XP_006254653.1 | 5 | 1704 | |||
| XM_006254593.2 | XP_006254655.1 | 5 | 1104 | |||
| XM_008772042.1 | XP_008770264.1 | 5 | 1704 | |||

Ammecr1l
Genomic Map