siRNA Details
| Gene Symbol: LOC103694178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772009.1 | XP_008770231.1 | 2 | 55 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103694178| Gene Symbol: LOC103694178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772009.1 | XP_008770231.1 | 2 | 55 | ||