siRNA Details
| Gene Symbol: Lama3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772008.1 | XP_008770230.1 | 61 | 8962 | ||
| GenBank mRNA | U61261.1 | - | 23 | 3318 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lama3| Gene Symbol: Lama3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772008.1 | XP_008770230.1 | 61 | 8962 | ||
| GenBank mRNA | U61261.1 | - | 23 | 3318 | ||