siRNA Details
Gene Symbol: Cdh16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012055.1 | NP_001012055.1 | 17, 18 | 2542 | ||
GenBank mRNA | BC081996.1 | - | 17, 18 | 2542 | 7130691 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdh16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012055.1 | NP_001012055.1 | 17, 18 | 2542 | ||
GenBank mRNA | BC081996.1 | - | 17, 18 | 2542 | 7130691 |