siRNA Details
| Gene Symbol: Ccdc79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772250.1 | XP_008770472.1 | 12 | 1401 | ||
| XM_008772251.1 | XP_008770473.1 | 12 | 1322 | |||
| XM_008772252.1 | XP_008770474.1 | 12 | 1609 | |||
| XM_008772253.1 | XP_008770475.1 | 11 | 1480 | |||
| XM_008772254.1 | XP_008770476.1 | 12 | 1612 | |||
| XM_008772255.1 | XP_008770477.1 | 11 | 1459 | |||
| XM_008772256.1 | XP_008770478.1 | 11 | 1456 | |||
| XM_008772257.1 | XP_008770479.1 | 12 | 1612 | |||

Ccdc79
Genomic Map