siRNA Details
| Gene Symbol: CDC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB003698.1 | - | 9 | 1118 | ||
| AB209337.1 | - | 6 | 3245 | |||
| AF005209.1 | - | 8 | 986 | |||
| AF015592.1 | - | 9 | 1131 | |||
| AK298963.1 | - | 8 | 1063 | |||
| AK312726.1 | - | 9 | 1207 | |||
| BC110526.1 | - | 9 | 1074 | 40034178 | ||
| BC110527.1 | - | 9 | 1108 | 40034179 | ||

CDC7
Genomic Map