siRNA Details
| Gene Symbol: Gins3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107408.1 | NP_001100878.1 | 1 | 121 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gins3| Gene Symbol: Gins3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107408.1 | NP_001100878.1 | 1 | 121 | ||