siRNA Details
| Gene Symbol: Cfap20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037978.1 | NP_001033067.1 | 4 | 476 | ||
| XM_006255083.1 | XP_006255145.1 | 4 | 562 | |||
| GenBank mRNA | BC099768.1 | - | 4 | 476 | 7932140 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfap20| Gene Symbol: Cfap20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037978.1 | NP_001033067.1 | 4 | 476 | ||
| XM_006255083.1 | XP_006255145.1 | 4 | 562 | |||
| GenBank mRNA | BC099768.1 | - | 4 | 476 | 7932140 | |