siRNA Details
Gene Symbol: Gpr114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107410.1 | NP_001100880.1 | 9 | 1044 | ||
XM_006255101.2 | XP_006255163.1 | 10 | 1117 | |||
XM_008772297.1 | XP_008770519.1 | 10 | 1102 | |||
XM_008772298.1 | XP_008770520.1 | 10 | 999 | |||
XM_008772299.1 | XP_008770521.1 | 10 | 1024 | |||
XM_008772300.1 | XP_008770522.1 | 9 | 1213 | |||
XM_008772301.1 | XP_008770523.1 | 10 | 1141 | |||
XM_008772302.1 | XP_008770524.1 | 10 | 1063 | |||
XM_008772303.1 | XP_008770525.1 | 10 | 1095 | |||
XM_008772304.1 | XP_008770526.1 | 10 | 1402 | |||
XM_008772305.1 | XP_008770527.1 | 10 | 1063 | |||
XM_008772306.1 | XP_008770528.1 | 10 | 1104 | |||
XM_008772307.1 | XP_008770529.1 | 10 | 1101 | |||
XM_008772308.1 | XP_008770530.1 | 10 | 1136 | |||
XM_008772309.1 | XP_008770531.1 | 10 | 1112 | |||
XM_008772310.1 | XP_008770532.1 | 10 | 1090 | |||
XM_008772311.1 | XP_008770533.1 | 10 | 1055 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
