siRNA Details
| Gene Symbol: Ces5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012056.1 | NP_001012056.1 | 3 | 447 | ||
| GenBank mRNA | AF479659.1 | - | 3 | 447 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ces5a| Gene Symbol: Ces5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012056.1 | NP_001012056.1 | 3 | 447 | ||
| GenBank mRNA | AF479659.1 | - | 3 | 447 | ||